Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCTATGTAAGTGTTGTATTTT[A/T]TTGTTGTTTGTTTTTTCCTAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601757 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX7 - peroxisomal biogenesis factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000288.3 | Intron | NP_000279.1 | ||||
XM_005267019.4 | Intron | XP_005267076.1 | ||||
XM_006715502.1 | Intron | XP_006715565.1 | ||||
XM_017010934.1 | Intron | XP_016866423.1 |