Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCGTGGATGGAGCAGGAGGGGT[C/T]AGAGTATTGGGACCGGGAGACACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLA-E - major histocompatibility complex, class I, E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005516.5 | 371 | Missense Mutation | TCA,TTA | S,L 78 | NP_005507.3 | |
XM_017010807.1 | 371 | Missense Mutation | TCA,TTA | S,L 119 | XP_016866296.1 | |
XM_017010808.1 | 371 | Missense Mutation | TCA,TTA | S,L 119 | XP_016866297.1 | |
XM_017010809.1 | 371 | Missense Mutation | TCA,TTA | S,L 78 | XP_016866298.1 |