Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATAGATATCATACCAGATGCT[A/C]TCTTCTTAATTAGCTTTAGGTAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606857 | ||||||||||||||||||||
Literature Links: |
GCLC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCLC - glutamate-cysteine ligase catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197115.1 | 1803 | Missense Mutation | AGA,ATA | R,I 527 | NP_001184044.1 | |
NM_001498.3 | 1803 | Missense Mutation | AGA,ATA | R,I 565 | NP_001489.1 | |
XM_017010749.1 | 1803 | Missense Mutation | AGA,ATA | R,I 328 | XP_016866238.1 |