Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAGTCTTTTCCACACTGGGGA[C/T]ACCCATAGTGTTTCTCCCGAGTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HCG16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCG16 - HLA complex group 16 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF311 - zinc finger protein 311 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010877.3 | 1797 | Missense Mutation | TAT,TGT | Y,C 445 | NP_001010877.2 | |
NM_001318534.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 353 | NP_001305463.1 | |
NM_001318535.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 353 | NP_001305464.1 | |
XM_005249019.3 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_005249076.1 | |
XM_006715067.2 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_006715130.1 | |
XM_011514510.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_011512812.1 | |
XM_011514511.2 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_011512813.1 | |
XM_011514512.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 470 | XP_011512814.1 | |
XM_011514513.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 469 | XP_011512815.1 | |
XM_011514514.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 453 | XP_011512816.1 | |
XM_011514516.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 397 | XP_011512818.1 | |
XM_011514517.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 353 | XP_011512819.1 | |
XM_011514518.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 353 | XP_011512820.1 | |
XM_017010756.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_016866245.1 | |
XM_017010757.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 478 | XP_016866246.1 | |
XM_017010758.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 445 | XP_016866247.1 | |
XM_017010759.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 445 | XP_016866248.1 | |
XM_017010760.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 397 | XP_016866249.1 | |
XM_017010761.1 | 1797 | Missense Mutation | TAT,TGT | Y,C 353 | XP_016866250.1 |