Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTATGTACAAAAGGTTCCTGAC[C/T]TTCATTTCATAACTTATTCTGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf222 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf222 - chromosome 6 open reading frame 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010903.4 | 3100 | UTR 3 | NP_001010903.3 | |||
XM_011514596.2 | 3100 | UTR 3 | XP_011512898.1 | |||
XM_011514597.2 | 3100 | UTR 3 | XP_011512899.1 | |||
XM_011514598.2 | 3100 | UTR 3 | XP_011512900.1 |
PNPLA1 - patatin like phospholipase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |