Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAATTCCTTTGGAAAGGCATA[A/T]GAAAATCTAGTTTTTACTGCAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNC - cyclin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013399.1 | 809 | Silent Mutation | TCA,TCT | S,S 36 | NP_001013417.1 | |
NM_005190.3 | 809 | Silent Mutation | TCA,TCT | S,S 121 | NP_005181.2 | |
XM_006715594.1 | 809 | Silent Mutation | TCA,TCT | S,S 121 | XP_006715657.1 | |
XM_011536232.2 | 809 | Silent Mutation | TCA,TCT | S,S 121 | XP_011534534.1 | |
XM_017011436.1 | 809 | Silent Mutation | TCA,TCT | S,S 121 | XP_016866925.1 |
TSTD3 - thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195131.1 | 809 | Intron | NP_001182060.1 | |||
XM_017010141.1 | 809 | Intron | XP_016865630.1 |