Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGGCAGGCAGAATGTCAACAC[C/T]GTATGCACCACTGAGTTAAGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR5V1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR5V1 - olfactory receptor family 5 subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030876.5 | 737 | Silent Mutation | ACA,ACG | T,T 160 | NP_110503.3 | |
XM_011514929.2 | 737 | Silent Mutation | ACA,ACG | T,T 160 | XP_011513231.1 | |
XM_011514930.2 | 737 | Silent Mutation | ACA,ACG | T,T 160 | XP_011513232.1 |