Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAGCAGAACTTCAAAGATATC[A/C]GATATCAAAATGACTTGAGCAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OGFRL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OGFRL1 - opioid growth factor receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324266.1 | 835 | Silent Mutation | AGA,CGA | R,R 114 | NP_001311195.1 | |
NM_024576.4 | 835 | Silent Mutation | AGA,CGA | R,R 113 | NP_078852.3 | |
XM_011536117.2 | 835 | UTR 5 | XP_011534419.1 | |||
XM_011536118.2 | 835 | UTR 5 | XP_011534420.1 | |||
XM_017011303.1 | 835 | Nonsense Mutation | AGA,CGA | R,R 128 | XP_016866792.1 | |
XM_017011304.1 | 835 | Silent Mutation | AGA,CGA | R,R 127 | XP_016866793.1 | |
XM_017011305.1 | 835 | Silent Mutation | AGA,CGA | R,R 51 | XP_016866794.1 | |
XM_017011306.1 | 835 | UTR 5 | XP_016866795.1 |