Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCCCACCAACGGGCACCATGGG[A/G]ATAGGGGCTCGCACTTGTTGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIVEP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIVEP2 - human immunodeficiency virus type I enhancer binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006734.3 | 7386 | Silent Mutation | ATC,ATT | I,I 2226 | NP_006725.3 | |
XM_017010805.1 | 7386 | Silent Mutation | ATC,ATT | I,I 2226 | XP_016866294.1 |