Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCTGGTCTCGGTTGTGGCGGC[C/T]GTGGGGCTCTTTGCTGGGCTCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616991 | ||||||||||||||||||||
Literature Links: |
MUC21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUC21 - mucin 21, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010909.3 | 1790 | Silent Mutation | GCC,GCT | A,A 491 | NP_001010909.2 | |
NM_001322370.1 | 1790 | Silent Mutation | NP_001309299.1 | |||
NM_001322371.1 | 1790 | Silent Mutation | NP_001309300.1 |