Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTTTCAGTTCCATGAAGGTGG[A/G]CTTCACACATGTGTTATTTGCTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607345 MIM: 603400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBE1 - tubulin epsilon 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016262.4 | 1347 | Missense Mutation | CCC,TCC | P,S 407 | NP_057346.1 | |
XM_011535875.2 | 1347 | Missense Mutation | CCC,TCC | P,S 332 | XP_011534177.1 | |
XM_017010918.1 | 1347 | Missense Mutation | CCC,TCC | P,S 363 | XP_016866407.1 |
WISP3 - WNT1 inducible signaling pathway protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |