Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCACTGCCCAGGAGAATGGAT[C/T]GCCCTTCGTCTAGGGCAAAAACATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604698 MIM: 616595 | ||||||||||||||||||||
Literature Links: |
AKAP12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP12 - A-kinase anchoring protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB2 - zinc finger and BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020861.2 | 1820 | Missense Mutation | CAA,CGA | Q,R 482 | NP_065912.1 | |
XM_005267076.2 | 1820 | Missense Mutation | CAA,CGA | Q,R 482 | XP_005267133.1 | |
XM_011536004.2 | 1820 | Missense Mutation | CAA,CGA | Q,R 482 | XP_011534306.1 | |
XM_017011137.1 | 1820 | Missense Mutation | CAA,CGA | Q,R 403 | XP_016866626.1 |