Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTGCATGAGATATTAAAGAAA[C/T]TGGATCCCAATGTGAATTCTGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MTFR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTFR2 - mitochondrial fission regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099286.2 | 1154 | Missense Mutation | ATT,GTT | I,V 310 | NP_001092756.1 | |
NM_001318738.1 | 1154 | Missense Mutation | ATT,GTT | I,V 267 | NP_001305667.1 | |
NM_138419.4 | 1154 | Missense Mutation | ATT,GTT | I,V 310 | NP_612428.2 | |
XM_011535410.2 | 1154 | Missense Mutation | ATT,GTT | I,V 267 | XP_011533712.1 | |
XM_011535412.2 | 1154 | Missense Mutation | ATT,GTT | I,V 267 | XP_011533714.1 | |
XM_011535413.2 | 1154 | Missense Mutation | ATT,GTT | I,V 267 | XP_011533715.1 |