Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGGCAGCCTCTGTCCATCTCGC[C/G]CAAGGAGCTCTGTCTGCAGTCGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124450 | ||||||||||||||||||||
Literature Links: |
DDO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDO - D-aspartate oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003649.2 | 786 | Missense Mutation | GCG,GGG | A,G 320 | NP_003640.2 | |
NM_004032.2 | 786 | Missense Mutation | GCG,GGG | A,G 261 | NP_004023.2 |