Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAAAATTCCACCCCTATAGTAA[C/T]ACCTGTATTCCTACTTCCTGTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF3 - PHD finger protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290259.1 | Intron | NP_001277188.1 | ||||
NM_001290260.1 | Intron | NP_001277189.1 | ||||
NM_015153.3 | Intron | NP_055968.1 | ||||
XM_005248701.3 | Intron | XP_005248758.1 | ||||
XM_005248702.3 | Intron | XP_005248759.1 | ||||
XM_005248703.3 | Intron | XP_005248760.1 | ||||
XM_006715426.3 | Intron | XP_006715489.1 | ||||
XM_011535648.2 | Intron | XP_011533950.1 | ||||
XM_017010626.1 | Intron | XP_016866115.1 |