Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCTCGTTGTGCTCCTTGGCCTT[C/T]TTCTGGCACATGGAGATCTGCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602623 | ||||||||||||||||||||
Literature Links: |
FKBP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKBP5 - FK506 binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145775.2 | 1346 | Silent Mutation | AAA,AAG | K,K 396 | NP_001139247.1 | |
NM_001145776.1 | 1346 | Silent Mutation | AAA,AAG | K,K 396 | NP_001139248.1 | |
NM_001145777.1 | 1346 | Intron | NP_001139249.1 | |||
NM_004117.3 | 1346 | Silent Mutation | AAA,AAG | K,K 396 | NP_004108.1 |