Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGACCTTTTATGGACTCCCTGGT[G/T]GGCACTGCTGCTGCTACAGGTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605310 MIM: 613525 MIM: 613526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCHCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCHCR1 - coiled-coil alpha-helical rod protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105563.1 | 2408 | Silent Mutation | CCA,CCC | P,P 788 | NP_001099033.1 | |
NM_001105564.1 | 2408 | Silent Mutation | CCA,CCC | P,P 824 | NP_001099034.1 | |
NM_019052.3 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | NP_061925.2 | |
XM_011514702.2 | 2408 | Silent Mutation | CCA,CCC | P,P 797 | XP_011513004.1 | |
XM_011514703.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_011513005.1 | |
XM_011514704.2 | 2408 | Silent Mutation | CCA,CCC | P,P 699 | XP_011513006.1 | |
XM_011514705.1 | 2408 | Silent Mutation | CCA,CCC | P,P 699 | XP_011513007.1 | |
XM_017010961.1 | 2408 | Silent Mutation | CCA,CCC | P,P 833 | XP_016866450.1 | |
XM_017010962.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866451.1 | |
XM_017010963.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866452.1 | |
XM_017010964.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866453.1 | |
XM_017010965.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866454.1 | |
XM_017010966.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866455.1 | |
XM_017010967.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866456.1 | |
XM_017010968.1 | 2408 | Silent Mutation | CCA,CCC | P,P 735 | XP_016866457.1 | |
XM_017010969.1 | 2408 | Silent Mutation | CCA,CCC | P,P 699 | XP_016866458.1 | |
XM_017010970.1 | 2408 | Silent Mutation | CCA,CCC | P,P 459 | XP_016866459.1 |
PSORS1C1 - psoriasis susceptibility 1 candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSORS1C2 - psoriasis susceptibility 1 candidate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |