Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGACTGGTTGAAAAAGTCTAAG[A/G]TTAAATGGATAAATACAATATAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TINAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TINAG - tubulointerstitial nephritis antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014464.3 | Intron | NP_055279.3 | ||||
XM_006715062.2 | Intron | XP_006715125.1 | ||||
XM_011514497.2 | Intron | XP_011512799.1 | ||||
XM_017010745.1 | Intron | XP_016866234.1 | ||||
XM_017010746.1 | Intron | XP_016866235.1 | ||||
XM_017010747.1 | Intron | XP_016866236.1 | ||||
XM_017010748.1 | Intron | XP_016866237.1 |