Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGCCTGCGCTCCTCCTGGAAGC[A/G]GATGAGGGCATCTCTCTGGTTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607129 MIM: 603498 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICAL1 - microtubule associated monooxygenase, calponin and LIM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159291.1 | 3493 | Missense Mutation | CGC,TGC | R,C 963 | NP_001152763.1 | |
NM_001286613.1 | 3493 | Missense Mutation | CGC,TGC | R,C 1068 | NP_001273542.1 | |
NM_022765.3 | 3493 | Missense Mutation | CGC,TGC | R,C 1049 | NP_073602.3 |
PPIL6 - peptidylprolyl isomerase like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPD2 - sphingomyelin phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003080.2 | 3493 | Intron | NP_003071.2 | |||
XM_005267109.1 | 3493 | Intron | XP_005267166.1 | |||
XM_011536079.1 | 3493 | Intron | XP_011534381.1 |