Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTATGGTTCAGCTGGAGAACA[A/G]AGGCCCTGTCCCGGGCATACATCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK5 - potassium two pore domain channel subfamily K member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003740.3 | 2046 | UTR 3 | NP_003731.1 | |||
XM_005249456.1 | 2046 | UTR 3 | XP_005249513.1 | |||
XM_006715235.2 | 2046 | UTR 3 | XP_006715298.1 |