Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGATGCTGTATTATCACTGCTTA[A/G]AGCAACAAGGCCCATATTCTTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186982 | ||||||||||||||||||||
Literature Links: |
TCP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCP11 - t-complex 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093728.2 | 1512 | Silent Mutation | CTA,TTA | L,L 413 | NP_001087197.1 | |
NM_001261817.1 | 1512 | Silent Mutation | CTA,TTA | L,L 408 | NP_001248746.1 | |
NM_001261818.1 | 1512 | Silent Mutation | CTA,TTA | L,L 367 | NP_001248747.1 | |
NM_001261819.1 | 1512 | Silent Mutation | CTA,TTA | L,L 362 | NP_001248748.1 | |
NM_001261820.1 | 1512 | Silent Mutation | CTA,TTA | L,L 337 | NP_001248749.1 | |
NM_001261821.1 | 1512 | Silent Mutation | CTA,TTA | L,L 337 | NP_001248750.1 | |
NM_018679.5 | 1512 | Silent Mutation | CTA,TTA | L,L 338 | NP_061149.1 | |
XM_005249339.2 | 1512 | Silent Mutation | CTA,TTA | L,L 259 | XP_005249396.1 | |
XM_011514830.1 | 1512 | Silent Mutation | CTA,TTA | L,L 516 | XP_011513132.1 | |
XM_011514831.1 | 1512 | Silent Mutation | CTA,TTA | L,L 259 | XP_011513133.1 | |
XM_011514832.1 | 1512 | Silent Mutation | CTA,TTA | L,L 259 | XP_011513134.1 |