Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTTGCTGCCAGGTTCAAGGA[A/T]TCTCTTAATTGTAGGGATATTACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605450 | ||||||||||||||||||||
Literature Links: |
GSTA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTA4 - glutathione S-transferase alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001512.3 | 739 | Missense Mutation | ATC,TTC | I,F 197 | NP_001503.1 | |
XM_005249035.3 | 739 | Missense Mutation | ATC,TTC | I,F 197 | XP_005249092.1 | |
XM_011514534.2 | 739 | Missense Mutation | ATC,TTC | I,F 160 | XP_011512836.1 | |
XM_011514535.2 | 739 | Missense Mutation | ATC,TTC | I,F 160 | XP_011512837.1 |