Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAGAAGACCGAGTCTTAGCCTT[A/G]GCGCGAGCTTTACCGCCTTGTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602795 MIM: 602803 MIM: 602819 MIM: 602829 | ||||||||||||||||||||
Literature Links: |
HIST1H2AB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST1H2AB - histone cluster 1, H2ab | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003513.2 | Intron | NP_003504.2 |
HIST1H2BB - histone cluster 1, H2bb | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3B - histone cluster 1, H3b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4B - histone cluster 1, H4b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |