Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTACAGACAGCCATCTACTCTGT[C/T]GTGGGGTGAGTACTTGTTTCTCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TSPO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPO2 - translocator protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010873.2 | 279 | Silent Mutation | GTC,GTT | V,V 56 | NP_001010873.1 | |
NM_001159726.1 | 279 | Silent Mutation | GTC,GTT | V,V 56 | NP_001153198.1 | |
XM_011514396.2 | 279 | Silent Mutation | GTC,GTT | V,V 56 | XP_011512698.1 | |
XM_011514397.2 | 279 | Silent Mutation | GTC,GTT | V,V 56 | XP_011512699.1 |
UNC5CL - unc-5 family C-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |