Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGTGCACTGGATGCCTTCACCT[A/G]TGGCCCTTCCCCAGCAACGGATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610774 MIM: 606817 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNPY3 - canopy FGF signaling regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNPY3-GNMT - CNPY3-GNMT readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCRA - pre T-cell antigen receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243168.1 | 308 | Missense Mutation | TAT,TGT | Y,C 76 | NP_001230097.1 | |
NM_001243169.1 | 308 | Missense Mutation | TAT,TGT | Y,C 51 | NP_001230098.1 | |
NM_001243170.1 | 308 | Intron | NP_001230099.1 | |||
NM_138296.2 | 308 | Missense Mutation | TAT,TGT | Y,C 76 | NP_612153.2 |