Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTGCATGTGGTTCCTTCTCTT[A/C]TTTCCTCAGACCACAAGATTCAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165020 | ||||||||||||||||||||
Literature Links: |
ROS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ROS1 - ROS proto-oncogene 1, receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002944.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2305 | NP_002935.2 | |
XM_006715548.3 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2300 | XP_006715611.1 | |
XM_011536049.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2315 | XP_011534351.1 | |
XM_011536050.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2314 | XP_011534352.1 | |
XM_011536051.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2306 | XP_011534353.1 | |
XM_011536052.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2301 | XP_011534354.1 | |
XM_011536053.2 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2257 | XP_011534355.1 | |
XM_011536054.2 | 7208 | Intron | XP_011534356.1 | |||
XM_011536055.2 | 7208 | Intron | XP_011534357.1 | |||
XM_011536056.2 | 7208 | Intron | XP_011534358.1 | |||
XM_011536057.2 | 7208 | Intron | XP_011534359.1 | |||
XM_011536058.2 | 7208 | Intron | XP_011534360.1 | |||
XM_017011172.1 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2292 | XP_016866661.1 | |
XM_017011173.1 | 7208 | Nonsense Mutation | GAA,TAA | E,* 2291 | XP_016866662.1 |