Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGAGGAGGAAGAAGAGGGCTG[C/T]GGTCATACACACTGTGTACGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606670 MIM: 607524 MIM: 607525 MIM: 615714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R11 - protein phosphatase 1 regulatory inhibitor subunit 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021959.2 | 468 | Intron | NP_068778.1 | |||
XM_006715174.3 | 468 | Silent Mutation | TGC,TGT | C,C 74 | XP_006715237.1 |
RNF39 - ring finger protein 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNRD1 - zinc ribbon domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNRD1ASP - zinc ribbon domain containing 1 antisense, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |