Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAACGACAGGACAAACAAAGCAC[A/G]CTAACCTAAAACACGGAAAGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611506 MIM: 609058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPQ - centromere protein Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018132.3 | 197 | Missense Mutation | ACT,GCT | T,A 58 | NP_060602.2 | |
XM_005249205.1 | 197 | Missense Mutation | ACT,GCT | T,A 58 | XP_005249262.1 | |
XM_011514723.2 | 197 | Missense Mutation | ACT,GCT | T,A 61 | XP_011513025.2 |
MUT - methylmalonyl-CoA mutase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |