Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAGCAGCATGTCCAGCTGAGCA[A/G]CCTCTTGGTACAGCTGGAAGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606998 MIM: 191130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLOT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLOT1 - flotillin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318875.1 | 1391 | Missense Mutation | GCT,GTT | A,V 303 | NP_001305804.1 | |
NM_005803.3 | 1391 | Missense Mutation | GCT,GTT | A,V 351 | NP_005794.1 | |
XM_005248780.3 | 1391 | Missense Mutation | GCT,GTT | A,V 351 | XP_005248837.1 | |
XM_006714947.3 | 1391 | Missense Mutation | GCT,GTT | A,V 298 | XP_006715010.1 | |
XM_017010157.1 | 1391 | Missense Mutation | GCT,GTT | A,V 347 | XP_016865646.1 | |
XM_017010158.1 | 1391 | Missense Mutation | GCT,GTT | A,V 314 | XP_016865647.1 |
TUBB - tubulin beta class I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |