Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGACATGGAGGATGCGGATAT[C/G]ACTCTGTGGACAGAGGCTGAGTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603423 | ||||||||||||||||||||
Literature Links: |
PRDM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRDM1 - PR domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198.3 | 159 | Missense Mutation | ATC,ATG | I,M 46 | NP_001189.2 | |
NM_182907.2 | 159 | Intron | NP_878911.1 | |||
XM_006715550.3 | 159 | Missense Mutation | ATC,ATG | I,M 60 | XP_006715613.1 | |
XM_011536062.2 | 159 | Missense Mutation | ATC,ATG | I,M 60 | XP_011534364.1 | |
XM_011536063.2 | 159 | Missense Mutation | ATC,ATG | I,M 10 | XP_011534365.1 | |
XM_011536064.2 | 159 | Intron | XP_011534366.1 | |||
XM_017011187.1 | 159 | Missense Mutation | ATC,ATG | I,M 10 | XP_016866676.1 |