Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTCCCATTGGTTTAAAATTTG[A/G]TAAGTGTATTTCTTTGCTGACAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STX7 - syntaxin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326578.1 | 4233 | UTR 3 | NP_001313507.1 | |||
NM_001326579.1 | 4233 | UTR 3 | NP_001313508.1 | |||
NM_001326580.1 | 4233 | Intron | NP_001313509.1 | |||
NM_003569.2 | 4233 | UTR 3 | NP_003560.2 | |||
XM_011536178.1 | 4233 | Intron | XP_011534480.1 |