Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACGACTGCTTCGTCAAGGTGCT[A/G]CGCGACCCCTCGCGGCCCTGGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612788 | ||||||||||||||||||||
Literature Links: |
FOXQ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXQ1 - forkhead box Q1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033260.3 | 808 | Silent Mutation | CTA,CTG | L,L 181 | NP_150285.3 |
LINC01394 - long intergenic non-protein coding RNA 1394 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107986554 - uncharacterized LOC107986554 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017011607.1 | 808 | Intron | XP_016867096.1 | |||
XM_017011608.1 | 808 | Intron | XP_016867097.1 | |||
XM_017011609.1 | 808 | Intron | XP_016867098.1 | |||
XM_017011610.1 | 808 | Intron | XP_016867099.1 | |||
XM_017011611.1 | 808 | Intron | XP_016867100.1 |