Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGGGCAGGAGGGAGCCTGGGA[C/G]GCCCAAGCCCTGGTGCTCAAGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142560 MIM: 609624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1G2-DDX39B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1G2-DDX39B - ATP6V1G2-DDX39B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX39B - DEAD-box helicase 39B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004640.6 | 332 | Intron | NP_004631.1 | |||
NM_080598.5 | 332 | Intron | NP_542165.1 |
MCCD1 - mitochondrial coiled-coil domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011700.2 | 332 | Missense Mutation | GAC,GAG | D,E 93 | NP_001011700.2 |
SNORD117 - small nucleolar RNA, C/D box 117 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |