Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOM - apolipoprotein M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C6orf47 - chromosome 6 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021184.3 | 1209 | Silent Mutation | CCA,CCC | P,P 128 | NP_067007.3 |
CSNK2B - casein kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPANK1 - G-patch domain and ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |