Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTCCTGGCCTGCCCTGGGTGGT[A/G]TGAACTACAAAGTTCTATTTGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC29A1 - solute carrier family 29 member 1 (Augustine blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078175.2 | Intron | NP_001071643.1 | ||||
NM_001078177.1 | Intron | NP_001071645.1 | ||||
NM_001304462.1 | Intron | NP_001291391.1 | ||||
NM_001304463.1 | Intron | NP_001291392.1 | ||||
NM_001304465.1 | Intron | NP_001291394.1 | ||||
NM_001304466.1 | Intron | NP_001291395.1 | ||||
XM_005248876.4 | Intron | XP_005248933.1 | ||||
XM_005248878.3 | Intron | XP_005248935.1 | ||||
XM_005248879.3 | Intron | XP_005248936.1 | ||||
XM_005248880.3 | Intron | XP_005248937.1 | ||||
XM_005248881.3 | Intron | XP_005248938.1 | ||||
XM_005248882.3 | Intron | XP_005248939.1 | ||||
XM_011514341.2 | Intron | XP_011512643.1 |