Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTGCAGTCCTGGATGGCGCCTC[C/T]GACTTGCAGCTTCCTTCCTGCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612509 | ||||||||||||||||||||
Literature Links: |
ABCC10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCC10 - ATP binding cassette subfamily C member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198934.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 69 | NP_001185863.1 | |
NM_033450.2 | 420 | Nonsense Mutation | CGA,TGA | R,* 26 | NP_258261.2 | |
XM_011514974.2 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_011513276.2 | |
XM_011514983.2 | 420 | Intron | XP_011513285.1 | |||
XM_011514985.2 | 420 | Intron | XP_011513287.1 | |||
XM_011514986.2 | 420 | Intron | XP_011513288.1 | |||
XM_017011445.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866934.1 | |
XM_017011446.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 69 | XP_016866935.1 | |
XM_017011447.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 69 | XP_016866936.1 | |
XM_017011448.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866937.1 | |
XM_017011449.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866938.1 | |
XM_017011450.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866939.1 | |
XM_017011451.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866940.1 | |
XM_017011452.1 | 420 | UTR 5 | XP_016866941.1 | |||
XM_017011453.1 | 420 | Nonsense Mutation | CGA,TGA | R,* 120 | XP_016866942.1 | |
XM_017011454.1 | 420 | Intron | XP_016866943.1 | |||
XM_017011455.1 | 420 | Intron | XP_016866944.1 | |||
XM_017011456.1 | 420 | Intron | XP_016866945.1 |
MIR6780B - microRNA 6780b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |