Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTTTATTTAGCCCACAATATT[C/T]AATCATGTAATCAATATTTTTAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN16 - zinc finger and SCAN domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320555.1 | Intron | NP_001307484.1 | ||||
NM_001320556.1 | Intron | NP_001307485.1 | ||||
NM_001320557.1 | Intron | NP_001307486.1 | ||||
NM_001320558.1 | Intron | NP_001307487.1 | ||||
NM_025231.2 | Intron | NP_079507.1 | ||||
XM_017011324.1 | Intron | XP_016866813.1 |
ZSCAN16-AS1 - ZSCAN16 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |