Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACCAGAGCCCAGGCCTAAAAAG[C/G]CCTCTGCAAAGAAGGGAGAGAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HMGN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGN4 - high mobility group nucleosomal binding domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006353.2 | 368 | Missense Mutation | CCC,GCC | P,A 44 | NP_006344.1 | |
XM_017010171.1 | 368 | Missense Mutation | CCC,GCC | P,A 44 | XP_016865660.1 | |
XM_017010172.1 | 368 | Missense Mutation | CCC,GCC | P,A 44 | XP_016865661.1 | |
XM_017010173.1 | 368 | Missense Mutation | CCC,GCC | P,A 44 | XP_016865662.1 |