Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGGGGGTGCTGCTGCCGCCGCT[A/G]TGCTTGGGGTTGCAATCTCGGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602937 | ||||||||||||||||||||
Literature Links: |
CITED2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CITED2 - Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168388.2 | 726 | Silent Mutation | CAC,CAT | H,H 160 | NP_001161860.1 | |
NM_001168389.2 | 726 | Silent Mutation | CAC,CAT | H,H 165 | NP_001161861.2 | |
NM_006079.4 | 726 | Silent Mutation | CAC,CAT | H,H 160 | NP_006070.2 |