Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTTGCCGCCCTGCTTGCCACG[C/T]CCCGACATGACGTAAAAAATTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602788 MIM: 602801 MIM: 602825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H2AK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H2AK - histone cluster 1, H2ak | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003510.2 | 9 | Silent Mutation | GGA,GGG | G,G 3 | NP_003501.1 |
HIST1H2BN - histone cluster 1, H2bn | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003520.3 | 9 | Intron | NP_003511.1 |
HIST1H4K - histone cluster 1, H4k | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |