Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTGCTGCTCTGAAAAATATTC[G/T]ATCAAAACGAAGTTTACAAGCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609269 MIM: 605764 | ||||||||||||||||||||
Literature Links: |
KIAA0319 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0319 - KIAA0319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDP2 - tyrosyl-DNA phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016614.2 | 974 | Silent Mutation | NP_057698.2 |