Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTAGGCATAGGAACCTTCCAA[C/T]TGGAACGTTCCAACATTCTCACGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYLK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYLK4 - myosin light chain kinase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249078.3 | 2465 | UTR 3 | XP_005249135.2 | |||
XM_005249079.2 | 2465 | UTR 3 | XP_005249136.1 | |||
XM_006715082.3 | 2465 | UTR 3 | XP_006715145.1 | |||
XM_011514567.2 | 2465 | UTR 3 | XP_011512869.1 | |||
XM_017010819.1 | 2465 | UTR 3 | XP_016866308.1 |