Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAAGATGCTGCAAAACGTGACTC[C/T]CCACAATAAGTACGTTTCCGCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD2 - bromodomain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113182.2 | 1324 | Missense Mutation | CCC,CTC | P,L 7 | NP_001106653.1 | |
NM_001199455.1 | 1324 | Missense Mutation | CCC,CTC | P,L 7 | NP_001186384.1 | |
NM_001199456.1 | 1324 | Intron | NP_001186385.1 | |||
NM_001291986.1 | 1324 | UTR 5 | NP_001278915.1 | |||
NM_005104.3 | 1324 | Missense Mutation | CCC,CTC | P,L 7 | NP_005095.1 |