Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGGCAGGGTAAGGAGTGTGAGT[C/T]GTCTCCAATGATAGGTAAAGTCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609244 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAET1E-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAET1E-AS1 - RAET1E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAET1G - retinoic acid early transcript 1G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001788.3 | 1055 | Missense Mutation | AAC,GAC | N,D 305 | NP_001001788.2 | |
XM_011535800.2 | 1055 | UTR 3 | XP_011534102.1 | |||
XM_017010831.1 | 1055 | UTR 3 | XP_016866320.1 |