Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGGTGTAGGAGCCACTGTCATC[A/G]TTCCAGACAGGTGGCTTGTTGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603170 MIM: 602280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEAD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEAD3 - TEA domain transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TULP1 - tubby like protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289395.1 | 1451 | Silent Mutation | AAC,AAT | N,N 416 | NP_001276324.1 | |
NM_003322.4 | 1451 | Silent Mutation | AAC,AAT | N,N 469 | NP_003313.3 |