Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCTGGTGTCCATGACCTCACA[C/T]TCGATCTGCATCATCTCCTCATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600984 MIM: 600985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF6B - activating transcription factor 6 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136153.1 | 1971 | Silent Mutation | GAA,GAG | E,E 643 | NP_001129625.1 | |
NM_004381.4 | 1971 | Silent Mutation | GAA,GAG | E,E 646 | NP_004372.3 |
TNXB - tenascin XB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |