Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGGACCTGGGGCAGTAGATGGA[A/G]TTGGATGTAGCCCAGAGGGAGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WASF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WASF1 - WAS protein family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024934.1 | 1567 | Missense Mutation | NP_001020105.1 | |||
NM_001024935.1 | 1567 | Missense Mutation | NP_001020106.1 | |||
NM_001024936.1 | 1567 | Missense Mutation | NP_001020107.1 | |||
NM_003931.2 | 1567 | Missense Mutation | NP_003922.1 | |||
XM_005267203.3 | 1567 | Missense Mutation | XP_005267260.1 | |||
XM_005267205.2 | 1567 | Missense Mutation | XP_005267262.1 | |||
XM_005267206.2 | 1567 | Missense Mutation | XP_005267263.1 | |||
XM_005267207.2 | 1567 | Missense Mutation | XP_005267264.1 | |||
XM_011536233.1 | 1567 | Missense Mutation | XP_011534535.1 | |||
XM_011536234.1 | 1567 | Missense Mutation | XP_011534536.1 | |||
XM_011536235.1 | 1567 | Missense Mutation | XP_011534537.1 | |||
XM_017011437.1 | 1567 | Missense Mutation | XP_016866926.1 | |||
XM_017011438.1 | 1567 | Missense Mutation | XP_016866927.1 | |||
XM_017011439.1 | 1567 | Missense Mutation | XP_016866928.1 | |||
XM_017011440.1 | 1567 | Missense Mutation | XP_016866929.1 | |||
XM_017011441.1 | 1567 | Missense Mutation | XP_016866930.1 | |||
XM_017011442.1 | 1567 | Missense Mutation | XP_016866931.1 | |||
XM_017011443.1 | 1567 | Missense Mutation | XP_016866932.1 | |||
XM_017011444.1 | 1567 | Missense Mutation | XP_016866933.1 |