Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTCTCTGACGACGGGGAGTACA[C/T]GTGCTTTTTCAGGGAGGATGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601610 | ||||||||||||||||||||
Literature Links: |
BTN1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTN1A1 - butyrophilin subfamily 1 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001732.2 | 388 | Missense Mutation | ACG,ATG | T,M 123 | NP_001723.2 | |
XM_005249340.3 | 388 | Missense Mutation | ACG,ATG | T,M 223 | XP_005249397.2 |