Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGCGGGTCCCGGGATGTGACC[C/G]GGGCTCTGCTTGTGGCTGCGGCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615353 | ||||||||||||||||||||
Literature Links: |
CCDC28A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC28A - coiled-coil domain containing 28A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015439.2 | 294 | Missense Mutation | CGG,GGG | R,G 47 | NP_056254.1 | |
XM_011535728.2 | 294 | Missense Mutation | CGG,GGG | R,G 47 | XP_011534030.1 |
GVQW2 - GVQW motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242740.1 | 294 | Intron | NP_001229669.1 |