Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCACTTGATGCCACTCTGTAC[A/G]ACCCGGTTCTTGCAGTTTCTCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604599 MIM: 606107 | ||||||||||||||||||||
Literature Links: |
EHMT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EHMT2 - euchromatic histone lysine methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289413.1 | 3187 | Silent Mutation | NP_001276342.1 | |||
NM_001318833.1 | 3187 | Silent Mutation | NP_001305762.1 | |||
NM_006709.4 | 3187 | Silent Mutation | NP_006700.3 | |||
NM_025256.6 | 3187 | Silent Mutation | NP_079532.5 | |||
XM_005248824.3 | 3187 | Silent Mutation | XP_005248881.1 | |||
XM_006714974.1 | 3187 | Silent Mutation | XP_006715037.1 | |||
XM_006714975.1 | 3187 | Silent Mutation | XP_006715038.1 | |||
XM_006714976.1 | 3187 | Silent Mutation | XP_006715039.1 | |||
XM_017010202.1 | 3187 | Silent Mutation | XP_016865691.1 |
SLC44A4 - solute carrier family 44 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |